Learn More
Research Products International Corp Traceable Jumbo Digital Thermometer, Small
SDP
Supplier:  Research Products International Corp 814148
Encompass
Ideal Digital Thermometer for monitoring temperatures in refrigerators, freezers, incubators, and water baths. Offered with a 3/16'' diameter x 3/4'' probe for immersion in tubes, heating blocks and water baths. Displays minimum, maximum, current probe temperature plus room temperature. Temperature range: -58: to 158°F (-50 to 70°C) with accuracy of +1°C. Additional features include: built-in alarm and sealed probe.
Supplied with battery, certificate of calibration and 10 foot cable.
Catalog No. 50-197-8021