Learn More
Cole-Parmer Traceable Monitoring Hygrometer, Simultaneously displays time-of-day, humidity and temperature
Catalog No. NC3788328 Shop All Cole-Parmer Products
Encompass_Preferred
The Traceable® Monitoring Hygrometer is designed for professionals who require high-accuracy humidity and temperature monitoring in laboratory, industrial, and cleanroom environments. Customers can choose between NIST-traceable certification or A2LA-accredited calibration, ensuring compliance with ISO, GMP, and FDA standards.
Catalog No. NC3788328 Supplier Cole-Parmer Supplier No. 2025030
The Traceable® Monitoring Hygrometer is designed for professionals who require high-accuracy humidity and temperature monitoring in laboratory, industrial, and cleanroom environments. Customers can choose between NIST-traceable certification or A2LA-accredited calibration, ensuring compliance with ISO, GMP, and FDA standards.