Learn More
Description
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied withindividual temperature chart, traceability information and Traceable™ Certificate
Specifications
Specifications
| Chemical Name or Material | Buffer Standards One-Shot pH 10.012(CRM) |
| pH | 10.012 |
| Quantity | 100mL; 6-pk. |
| Packaging | 100 mL One-Shot (6/pk) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.