Learn More
Research Products International Corp Traceable pH Buffer Standards, 10.012 pH Value, 6/pk
SDP
Catalog No. 501978443
Encompass
Traceable pH Buffers provide the most precise accuracy available, ± 0.010 pH @ 25°C. These buffers have a Certified Reference Materials Classification and meet all Federal, State and local requirements to provide exact calibration results with all pH meters. Each buffer is supplied with an individual and serialized Traceable Certificate of Calibration. Convenient 100ml single use containers eliminate the need for long term storage.
Catalog No. 50-197-8443 Supplier Research Products International Corp Supplier No. 844289
Traceable pH Buffers provide the most precise accuracy available, ± 0.010 pH @ 25°C. These buffers have a Certified Reference Materials Classification and meet all Federal, State and local requirements to provide exact calibration results with all pH meters. Each buffer is supplied with an individual and serialized Traceable Certificate of Calibration. Convenient 100ml single use containers eliminate the need for long term storage.