Learn More
Marketlab Traceable Precision Monitoring Thermohygrometer
Catalog No. 22505048
Change view
Click to view available options
Type:
Electronic Thermohygrometer
Catalog No. Type
22-505-048 Electronic Thermohygrometer
1 options
Catalog No. 22-505-048 Supplier Marketlab Supplier No. 62190
Add to Cart
Add to Cart
Combines thermometer, clock, and humidity monitoring in one device.
  • Simultaneously displays time, current temperature, humidity, and a seven-day min/max graph
  • Easily scroll through the history for temperature or humidity min/max for the previous seven days
  • Internal sensors make it ideal for use in hoods, storerooms, cleanrooms, incubators, drying chambers, and environmental cabinets
  • Switchable between Fahrenheit and Celsius
  • 12/24 hour clock
  • Resolution: 0.1°
Specifications
Type Electronic Thermohygrometer
Humidity Range 0 to 90%