Learn More
Research Products International Corp Traceable Refrigerator Thermometer
SDP
Catalog No. 50999354
Encompass
Economical refrigerator thermometer has a temperature range from -9.9°C to 55.0°C (14.2°F to 131.0°F). Readings can be displayed in either °C or °F. Accuracy is +/-0.5°C with a resolution of 0.1°C. Display updates every thirty seconds for convenient temperature monitoring. Compact size allows placement in many locations.
Supplied with battery, suction cup mount and calibration certificate.
Catalog No. 50-999-354 Supplier Research Products International Corp Supplier No. 810011
Economical refrigerator thermometer has a temperature range from -9.9°C to 55.0°C (14.2°F to 131.0°F). Readings can be displayed in either °C or °F. Accuracy is +/-0.5°C with a resolution of 0.1°C. Display updates every thirty seconds for convenient temperature monitoring. Compact size allows placement in many locations.
Supplied with battery, suction cup mount and calibration certificate.