Learn More
Cole-Parmer Traceable Memory-Loc™ Datalogging Thermometer with Calibration; 1 Stainless Steel Probe
Catalog No. NC2119083 Shop All Cole-Parmer Products
Encompass_Preferred
Easily monitor temperatures overnight or for any time period—stores over 1 million temperature observations. Datalogger will stop recording when storage is full and will not overwrite recorded data. Memory can be cleared after downloading data and alarm events. Set high and low alarms in 1° increments; alarm sounds when temperature rises above or falls below set points. The alarm tracking feature stores data for up to 10 different alarm events detailing when the alarm state occurred, as well as when the unit returned within range. Alarm state indicator include audio alerts and flashing LCD segments. Transfer data recorded in a CSV format to any computer via a USB flash drive—no software is required. Unit continues to record data while downloading data. Sampling rate is 5 sec (10 sec for models 98768-55, -56, -59, and -60).
This item is not returnable. View return policy
Catalog No. NC2119083 Supplier Cole-Parmer Supplier No. 9446044
Easily monitor temperatures overnight or for any time period—stores over 1 million temperature observations. Datalogger will stop recording when storage is full and will not overwrite recorded data. Memory can be cleared after downloading data and alarm events. Set high and low alarms in 1° increments; alarm sounds when temperature rises above or falls below set points. The alarm tracking feature stores data for up to 10 different alarm events detailing when the alarm state occurred, as well as when the unit returned within range. Alarm state indicator include audio alerts and flashing LCD segments. Transfer data recorded in a CSV format to any computer via a USB flash drive—no software is required. Unit continues to record data while downloading data. Sampling rate is 5 sec (10 sec for models 98768-55, -56, -59, and -60).