Learn More
Research Products International Corp Traceable Triple Display Timer, Chemical Resistant ABS Plastic Case
SDP
Catalog No. 50550351
Encompass
User friendly timer displays three different countdown/alarm times simultaneously. Three unique channels may be started individually or concurrently and programmed to count down from 99 hours, 59 minutes, 59 seconds to 1 second. When zero is reached, and alarm sounds, "Time's Up" is displayed and the timer begins counting up.
Memories return the display to the previously programmed time. May also be used as three stopwatches or in any combination of up/down timing. Display has a time of day clock.Supplied with a 1 1/2 year silver oxide battery. To assure accuracy an individually serial numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST. Quartz crystal timing accuracy is 0.01%.
Supplied with a magnetic back, easel stand, Traceable Certificate and a 1 1/2 year silver oxide battery. Chemical resistant ABS plastic case.
Catalog No. 50-550-351 Supplier Research Products International Corp Supplier No. 810014
User friendly timer displays three different countdown/alarm times simultaneously. Three unique channels may be started individually or concurrently and programmed to count down from 99 hours, 59 minutes, 59 seconds to 1 second. When zero is reached, and alarm sounds, "Time's Up" is displayed and the timer begins counting up.
Memories return the display to the previously programmed time. May also be used as three stopwatches or in any combination of up/down timing. Display has a time of day clock.Supplied with a 1 1/2 year silver oxide battery. To assure accuracy an individually serial numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST. Quartz crystal timing accuracy is 0.01%.
Supplied with a magnetic back, easel stand, Traceable Certificate and a 1 1/2 year silver oxide battery. Chemical resistant ABS plastic case.