Learn More
Fisherbrand™ TraceableLIVE™ Subscription Coupon Codes
Catalog No. 15710068 Shop All Fisherbrand Products
Change view
Click to view available options
Description:
1 Year Premium Subscription Coupon Code
1 Year Standard Subscription Coupon Code
2 Year Premium Subscription Coupon Code
2 Year Standard Subscription Coupon Code
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
15-710-068 2 Year Premium Subscription Coupon Code
15-710-065 1 Year Standard Subscription Coupon Code
15-710-066 2 Year Standard Subscription Coupon Code
15-710-067 1 Year Premium Subscription Coupon Code
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-710-068
Add to Cart
Add to Cart
TraceableLIVE™ Subscription Coupon Codes with simplified cloud monitoring for critical environments.
FREE - Try TraceableLIVE for free. Get Premium benefits for 30 days.
  • E-mail and push alarms
  • 1 user only
  • 1 month rolling data storage
  • Up to 3 devices can be added to the account
  • Calibration due and other notifications
Standard - Equip your team to protect your sensitive items and meet most national and local regulations.
  • E-mail and push alarms
  • Unlimited users
  • Unlimited data storage
  • No limit to devices that can be added to account (each device must have subscription)
  • Calibration due and other notifications
  • Reports: PDF, Secured PDF, CSV and VFC
  • Pre-loaded templates set up for monitoring Spex™ chemical products
Premium -Future-proof your vaccine and sample monitoring for ever-changing compliance requirements.
  • E-mail and push alarms
  • Unlimited users
  • Unlimited data storage
  • Apple Watch compatibility
  • SMS text notifications included
  • Voice notification escalation
  • No limit to devices that can be added to account (each device must have subscription)
  • Create Recurring Reports: PDF, Secured PDF, CSV and VFC
  • Create your own monitoring and notification templates
  • Unlimited templates
  • Pre-loaded templates set up for monitoring Spex chemical products
  • 21 CFR Part 11 compliance**
**Meets 21 CFR Part 11 North American Compliance and Annex 11 for Europe
Specifications
For Use With (Equipment) Compatible Traceable Devices
Certifications/Compliance 21 CFR Part 11 Compliance
Description 2 Year Premium Subscription Coupon Code
Product Type Coupon Code
For Use With (Application) Cloud Monitoring
Includes Card with Unique Coupon Code and QR Code
WARNING: Cancer - www.P65Warnings.ca.gov