Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 2,4-D Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018953 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8953 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8953 Supplier MilliporeSigma Supelco Supplier No. 76514100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 94-75-7
Vapor Pressure 0.4 mmHg (160°C)
Physical Form Neat
Linear Formula Cl2C6H3 OCH2 CO2 H
MDL Number MFCD00004300
Melting Point 136°C to 140°C
Synonym 2,4-Dichlorophenoxyacetic acid; 2,4-D
Type Certified Reference Material
Quantity 100 mg
Formula Weight 221.04
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.