Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 2,4-Dichlorophenol Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018940 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8940 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-8940 Supplier MilliporeSigma Supelco Supplier No. 07846100MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Product Line TraceCERT
CAS 120-83-2
Flash Point 114°C
Linear Formula Cl2C6H3 OH
MDL Number MFCD00002169
Melting Point 42°C to 43°C
Boiling Point 209°C to 210°C
Type Certified Reference Material
Quantity 100 mg
Formula Weight 163.00
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.