Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 2-Phenylethylamine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013244 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3244 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3244 Supplier MilliporeSigma Supelco Supplier No. 4268150MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 156-28-5
Linear Formula C6H5 CH2 CH2 NH2 · HCl
MDL Number MFCD00012893
Melting Point 220°C to 222°C
Synonym beta-Phenylethylamine hydrochloride; Phenethylamine hydrochloride
Type Certified Reference Material
Quantity 50 mg
Formula Weight 157.64
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.