Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 4,4-Diaminodiphenylmethane Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018649 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8649 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8649 Supplier MilliporeSigma Supelco Supplier No. 1813950MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 101-77-9
Linear Formula C13H14N2
MDL Number MFCD00007914
Melting Point 88°C to 92°C
Synonym 4,4 ′-Methylenedianiline; MDA
Type Certified Reference Material
Quantity 50 mg
Formula Weight 198.26
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.