Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Aflatoxin Mix in Methanol (Varied Conc.), MilliporeSigma™ Supelco™

Catalog No. 111017155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7155 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7155 Supplier MilliporeSigma Supelco Supplier No. CRM46304
Add to Cart
Add to Cart

Specifications

Packaging Ampule
CAS 67-56-1
Product Line TraceCERT
Flash Point 11°C - closed cup
Concentration Varied Conc.
Synonym Aflatoxin Mix
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.