Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Anthracene certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019049 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9049 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9049 Supplier MilliporeSigma Supelco Supplier No. 07671100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
Ignition Point 1004°F
CAS 120-12-7
Vapor Pressure 1 mmHg (145°C)
Vapor Density 6.15 (vs. air)
Flash Point 121°C
Physical Form Powder/Crystals
Linear Formula C14H10
MDL Number MFCD00001240
Melting Point 210°C to 215°C
Boiling Point 340°C
Synonym Anthraxcene; Paranaphthalene
Solubility Soluble in hydronaphthalenes
Type Certified Reference Material
Quantity 100 mg
Formula Weight 178.23
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.