Learn More
TraceCERT™ Aramite, 2,000 μg/mL in Methylene Chloride, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| CAS | 140-57-8 |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL in methylene chloride |
| Linear Formula | C15H23 ClO4 S |
| Synonym | Aramite solution |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 334.86 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.