Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Atrazine certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018992 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8992 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8992 Supplier MilliporeSigma Supelco Supplier No. 9093550MG
Add to Cart
Add to Cart

Atrazine is a commonly used herbicide, which belongs to the triazine class of herbicides.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 1912-24-9
Physical Form Neat
Linear Formula C8H14 ClN5
MDL Number MFCD00041810
Synonym 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 215.68
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.