Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Benzo[a]pyrene Solution, 1,000 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111017902 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7902 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7902 Supplier MilliporeSigma Supelco Supplier No. CRM40071
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 50-32-8
Flash Point -17.0°C - Closed Cup
Linear Formula C20H12
MDL Number MFCD00003602
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 252.31
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in acetone
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.