Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Benzo[k]fluoranthene Solution, 1,000 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111020430 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0430 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0430 Supplier MilliporeSigma Supelco Supplier No. 40073
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 207-08-9
Flash Point -17°C
Linear Formula C20H12
MDL Number MFCD00046287
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 252.31
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in acetone
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.