Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ BisphenolA certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019055 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9055 Supplier MilliporeSigma Supelco Supplier No. 42088100MG
Add to Cart
Add to Cart

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 80-05-7
Flash Point 227°C
Physical Form Neat
Linear Formula (CH3)2 C(C6H4 OH)2
MDL Number MFCD00002366
Melting Point 158°C to 159°C
Boiling Point 220°C (4 mmHg)
Synonym 2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol
Type Certified Reference Material
Quantity 100 mg
Formula Weight 228.29
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.