Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ cis-11-Vaccenic Acid Methyl Ester, 10 mg/mL in Heptane, MilliporeSigma™ Supelco™

Catalog No. 111020339 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0339 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0339 Supplier MilliporeSigma Supelco Supplier No. CRM46904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 1937-63-9
Product Line TraceCERT
Flash Point -3.99°C - closed cup
Shipping Condition Ambient
Concentration 10 mg/mL in heptane
Linear Formula CH3(CH2)5 CH=CH(CH2)9 CO2 CH3
Synonym cis-11-Octadecenoic methyl ester; cis-Vaccenic acid methyl ester
Type Certified Reference Material
Quantity 1 mL
Formula Weight 296.49
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.