Learn More
TraceCERT™ cis-6-Octadecenoic acid methyl ester, 10 mg/mL in heptane, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 2777-58-4 |
| Flash Point | -4°C |
| Physical Form | Liquid |
| Synonym | Petroselinic acid methyl ester |
| Type | Certified Reference Material |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 10 mg/mL in heptane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.