Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Clothianidin, MilliporeSigma™ Supelco™

Catalog No. 111018432 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8432 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8432 Supplier MilliporeSigma Supelco Supplier No. 6896550MG
Add to Cart
Add to Cart

Find more information here: Neonicotinoids

TRUSTED_SUSTAINABILITY

Specifications

CAS 210880-92-5
Product Line TraceCERT
Linear Formula C6H8 ClN5O2 S
MDL Number MFCD06200753
Synonym (E)-1-(2-Chloro-5-thiazolylmethyl)-3-methyl-2-nitroguanidine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 249.68
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.