Learn More
TraceCERT™ DBA Isocyanate Monomers Mix, Certified Reference Material, in Acetone (varied concentration), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Synonym | Dibutylamine Isocyanate Monomers Mix |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | ICA-DBA DERIVATIVE ; MIC-DBA DERIVATIVE; EIC-DBA DERIVATIVE; PIC-DBA DERIVATIVE; PHI-DBA DERIVATIVE; 1,6-HDI-DBA DERIVATIVE; 2,4-TDI-DBA DERIVATIVE; 2,6-TDI-DBA DERIVATIVE ; 4,4-MDI-DBA DERIVATIVE ; IPDI-DBA DERIVATIVE ISOMER 1; IPDI-DBA DERIVATIVE ISOMER 2 |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.