Learn More
TraceCERT™ Decafluorobiphenyl Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Chemical Name or Material | Decafluorobiphenyl |
| Packaging | Ampule |
| CAS | 434-90-2 |
| Product Line | TraceCERT |
| Concentration | 2000 μg/mL |
| Technique | HPLC |
| Certifications/Compliance | ISO 17034 |
| Type | Organic Pollutant Standard |
| Solvent or Matrix | Dichloromethane |
| Molecular Weight (g/mol) | 334.09 |
| Show More |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.