Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dibutyl sulfide certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018838 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8838 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8838 Supplier MilliporeSigma Supelco Supplier No. 76693100MG
Add to Cart
Add to Cart

Specifications

Product Line TraceCERT
CAS 544-40-1
Vapor Pressure 5.17 mmHg (37.7°C)
Vapor Density 5.07 (vs. air)
Density 0.838 g/mL (at 25°C)
Flash Point 77°C - Closed Cup
Linear Formula CH3(CH2)3 S(CH2)3 CH3
MDL Number MFCD00009468
Melting Point -76°C
Boiling Point 188°C to 189°C
Synonym 1,1 ′-Thiobisbutane; Butyl sulfide; NSC 8460
Type Certified Reference Material
Quantity 100 mg
Formula Weight 146.29
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.