Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Digoxin certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019053 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9053 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9053 Supplier MilliporeSigma Supelco Supplier No. 0459950MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 20830-75-5
Physical Form Powder
Linear Formula C41H64O14
MDL Number MFCD00003674
Synonym 12 beta-Hydroxydigitoxin
Type Certified Reference Material
Quantity 50 mg
Formula Weight 780.94
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.