Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dimethyl terephthalate Standard, MilliporeSigma™ Supelco™

Catalog No. 111012751 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2751 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2751 Supplier MilliporeSigma Supelco Supplier No. 070381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Environmental, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
Ignition Point 1058°F
CAS 120-61-6
Vapor Pressure 1.15 mmHg (93°C)
Vapor Density 1.04 (vs. air)
Flash Point 151°C
Linear Formula C6H4-1,4-(CO2 CH3)2
MDL Number MFCD00008440
Melting Point 139°C to 141°C
Type Certified Reference Material
Quantity 1 g
Formula Weight 194.18
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.