Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ DL-α-Tocopherol acetate, MilliporeSigma™ Supelco™

Catalog No. 111015481 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5481 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5481 Supplier MilliporeSigma Supelco Supplier No. 9235950MG
Add to Cart
Add to Cart

DL-α-Tocopherol acetate is a stable ester form of vitamin E, widely used in the formulation of cosmetics for the prevention or correction of skin damage.

Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 7695-91-2
Density 0.96 g/mL (at 20°C)
Physical Form Viscous Liquid
Linear Formula C31H52O3
MDL Number MFCD00072042
Synonym Vitamin E acetate; all-rac-alpha-Tocopheryl acetate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 472.74
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.