Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dodecane certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019173 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9173 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9173 Supplier MilliporeSigma Supelco Supplier No. 92064100MG
Add to Cart
Add to Cart

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
Ignition Point 401°F
CAS 112-40-3
Vapor Pressure 1 mmHg (47.8°C)
Vapor Density 5.96 (vs. air)
Density 0.75 g/mL (at 25°C)
Flash Point 71°C
Linear Formula CH3(CH2)10 CH3
MDL Number MFCD00008969
Melting Point -9.6°C
Boiling Point 215°C to 217°C
Type Certified Reference Material
Quantity 100 mg
Formula Weight 170.33
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.