Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ EPA 8270 Semivolatile Internal Standard Mix, 2,000 μg/mL, MilliporeSigma™ Supelco™

Catalog No. 111017765 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7765 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-7765 Supplier MilliporeSigma Supelco Supplier No. 46866U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 75-09-2
Type Organic Pollutant Standard
Quantity 25 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL each component in methylene chloride

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.