Learn More
TraceCERT™ Eugenol, 2000 μg/mL, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| CAS | 97-53-0 |
| Flash Point | 9.7°C (Closed Cup) |
| Concentration | 2000 μg/mL in methanol |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.