Learn More
TraceCERT™ F.A.M.E. Mix C8 - C24, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111020894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
11-102-0894 100 mg
1 options
Catalog No. 11-102-0894 Supplier MilliporeSigma Supelco Supplier No. CRM18918
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Product Line TraceCERT
Quantity 100 mg
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Methyl arachidate 8 wt. %; Methyl behenate 8 wt. %; Methyl decanoate 8 wt. %; Methyl cis-13-docosenoate 5 wt. %; Methyl dodecanoate 8 wt. %; Methyl linoleate; Methyl linolenate 5 wt. %; Methyl myristate 8 wt. %; Methyl octanoate 8 wt. %; Methyl oleate 5 wt. %; Methyl palmitate 11 wt. %; Methyl palmitoleate 5 wt. %; Methyl stearate 8 wt. %; Methyl tetracosanoate 8 wt. %
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC