Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ F.A.M.E. Mix GLC-10 (Neat), MilliporeSigma™ Supelco™

Catalog No. 111005049 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-5049 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5049 Supplier MilliporeSigma Supelco Supplier No. CRM1891
Add to Cart
Add to Cart

This certified reference material is useful for determining relative retention times and approximating response factors.

TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
Shipping Condition Dry Ice
Physical Form Liquid
Type Certified Reference Material
Quantity 100 mg
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.