Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ F.A.M.E. Mix RM-6 (Neat), MilliporeSigma™ Supelco™

Catalog No. 111002704 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2704 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2704 Supplier MilliporeSigma Supelco Supplier No. O76311AMP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists' Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 1120-25-8
Product Line TraceCERT
Shipping Condition Dry Ice
Physical Form Neat
Synonym AOCS Reference Mixtures; AOCS No. 6 (RM-6); Oil Reference Standard, AOCS No. 6; Oil Reference Standard, AOCS
Type Certified Reference Material
Quantity 100 mg
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.