Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Fragrance Allergen Mix A2, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022428 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2428 2 mL
11-102-2429 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2428 Supplier MilliporeSigma Supelco Supplier No. 165582ML
Add to Cart
Add to Cart

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

  • 40 Components in Tert-Butyl Methyl Ether
TRUSTED_SUSTAINABILITY

Specifications

Flash Point -33°C (Closed Cup)
Quantity 2 mL
UN Number UN 2398 3/PGII
Grade Trace CERT

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.