Learn More
TraceCERT™ Fumaric Acid Solution, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 110-17-8 |
| Molecular Formula | C4H4O4 |
| Linear Formula | C4H4O4 |
| Quantity | 5 x 1 mL |
| MDL Number | MFCD00002700 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 116.07 |
| Grade | Trace CERT |
| Concentration or Composition (by Analyte or Components) | 1 mg/g in D2O |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.