Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Glufosinate-ammonium, MilliporeSigma™ Supelco™

Catalog No. 111015489 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5489 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5489 Supplier MilliporeSigma Supelco Supplier No. 49677100MG
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 77182-82-2
Linear Formula C5H15N2O4 P
MDL Number MFCD00055562
Melting Point 215°C
Synonym 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt; DL-Phosphinothricin
Type Certified Reference Material
Quantity 100 mg
Formula Weight 198.16
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.