Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Glyphosate certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019226 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9226 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9226 Supplier MilliporeSigma Supelco Supplier No. 89432100MG
Add to Cart
Add to Cart

Specifications

Product Line TraceCERT
CAS 1071-83-6
Linear Formula (HO)2 P(O)CH2 NHCH2 CO2 H
MDL Number MFCD00055350
Melting Point 230°C
Synonym N-(Phosphonomethyl)glycine; Glyphosate
Type Certified Reference Material
Quantity 100 mg
Formula Weight 169.07
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.