Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ L-Ascorbic Acid, 99.1%, MilliporeSigma™ Supelco™

Catalog No. 111014383 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4383 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4383 Supplier MilliporeSigma Supelco Supplier No. 57803100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-81-7
Product Line TraceCERT
Physical Form Neat
Linear Formula C6H8O6
MDL Number MFCD00064328
pH 1.0 to 2.5 (25°C, 176 g/L aq. soln.)
Melting Point 190°C to 194°C
Synonym L-Threoascorbic acid; Antiscorbutic factor; Vitamin C
Type Certified Reference Material
Quantity 100 mg
Formula Weight 176.12
Percent Purity 99.1%
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.