Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ L-Glutamine, MilliporeSigma™ Supelco™

Catalog No. 111014661 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4661 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4661 Supplier MilliporeSigma Supelco Supplier No. 76523100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 56-85-9
Physical Form Neat
Linear Formula H2 NCOCH2 CH2 CH(NH2)CO2 H
MDL Number MFCD00008044
Melting Point 185°C
Synonym (S)-2,5-Diamino-5-oxopentanoic acid; L-Glutamic acid 5-amide; Levoglutamide
Type Certified Reference Material
Quantity 100 mg
Formula Weight 146.14
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.