Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ L-Lysine monohydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111015089 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5089 Supplier MilliporeSigma Supelco Supplier No. 67448100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Lysine monohydrochloride is generally used as a food and feed supplement.It can also find applications in medicine and pharmaceutical industries.L-Lysine monohydrochloride can be produced via sugar fermentation by modified microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

CAS 657-27-2
Product Line TraceCERT
Physical Form Neat
Linear Formula H2 N(CH2)4 CH(NH2)CO2 H·HCl
MDL Number MFCD00064564
Melting Point 263°C
Synonym (S)-2,6-Diaminohexanoic acid monohydrochloride
Type Certified Reference Material
Quantity 100 mg
Formula Weight 182.65
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.