Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Methamidophos certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018425 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8425 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8425 Supplier MilliporeSigma Supelco Supplier No. 8993350MG
Add to Cart
Add to Cart

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 10265-92-6
Flash Point 212°C - Closed Cup
Linear Formula C2H8 NO2 PS
MDL Number MFCD00041808
Synonym O,S-Dimethyl phosphoramidothioate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 141.13
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.