Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Methyl cis-5,8,11,14-eicosatetraenoate Solution, 10 mg/mL in heptane, MilliporeSigma™ Supelco™

Catalog No. 111017127 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7127 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7127 Supplier MilliporeSigma Supelco Supplier No. 47572U
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 2566-89-4
Physical Form Liquid
Type Certified Reference Material
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 10 mg/mL in heptane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.