Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Methyl Ethanesulfonate, MilliporeSigma™ Supelco™

Catalog No. 111013410 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
120 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3410 120 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3410 Supplier MilliporeSigma Supelco Supplier No. 50490120MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1912-28-3
Product Line TraceCERT
Flash Point 83°C
Physical Form Neat
Linear Formula C2H5 SO3 CH3
MDL Number MFCD01728625
Synonym Ethanesulfonic acid methyl ester
Type Certified Reference Material
Quantity 120 mg
Formula Weight 124.16
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.