Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Methyl myristate, MilliporeSigma™ Supelco™

Catalog No. 111012827 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2827 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2827 Supplier MilliporeSigma Supelco Supplier No. 55791100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 124-10-7
Density 0.855 g/mL (at 25°C)
Flash Point 112°C
Physical Form Neat
Linear Formula CH3(CH2)12 COOCH3
MDL Number MFCD00008983
Melting Point 18°C
Boiling Point 323°C
Synonym Methyl tetradecanoate; Myristic acid methyl ester
Type Certified Reference Material
Quantity 100 mg
Formula Weight 242.40
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.