Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Methyl Palmitate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022615 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2615 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2615 Supplier MilliporeSigma Supelco Supplier No. 50374100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

TRUSTED_SUSTAINABILITY

Specifications

CAS 112-39-0
Melting Point 32°C to 35°C (literature)
Density 0.852 g/mL (at 25°C (literature))
Boiling Point 185°C (10 mmHg) (literature)
Flash Point 113°C (Closed Cup)
Molecular Formula C17H34O2
Linear Formula CH3(CH2)14 CO2CH3
Refractive Index n20/D 1.4512 (literature)
MDL Number MFCD00008994
Quantity 100 mg
Synonym n-Hexadecanoic acid methyl ester; Methyl hexadecanoate; Palmitic acid methyl ester
Formula Weight 270.45
Grade Trace CERT
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.