Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Octanoic acid certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019264 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9264 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9264 Supplier MilliporeSigma Supelco Supplier No. 0775450MG
Only null left
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 124-07-2
Vapor Pressure 1 mmHg (78°C)
Vapor Density 5 (vs. air)
Density 0.91 g/mL (at 25°C)
Physical Form Liquid
Linear Formula CH3(CH2)6 COOH
MDL Number MFCD00004429
Melting Point 15°C to 17°C
Boiling Point 237°C
Synonym Acid C8; Caprylic acid
Type Certified Reference Material
Quantity 50 mg
Formula Weight 144.21
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.