Learn More
TraceCERT™ Pinnatoxin F Solution, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1227167-70-5 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 20 μg/g (In 0.1% Acetic Acid in 50% Acetonitrile, Nominal Concentration) |
| Grade | Trace CERT |
| Quantity | 0.5 mL |
| Flash Point | 2°C (Closed Cup) |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.