Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Potassium phosphate monobasic, 31P-qNMR Standard, MilliporeSigma™ Supelco™

Catalog No. 111015517 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5517 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5517 Supplier MilliporeSigma Supelco Supplier No. 922141G
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 7778-77-0
Density 2.338 g/mL (at 25°C)
Linear Formula KH2 PO4
MDL Number MFCD00011401
Melting Point 252.6°C
Synonym Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
Type Certified Reference Material
Quantity 1 g
Formula Weight 136.09
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.